Anti-RELB Mouse Monoclonal Antibody [clone: 315206]

Supplier: R & D Systems
MAB2698
220020-038EA 594.37 USD
220020-038
Anti-RELB Mouse Monoclonal Antibody [clone: 315206]
Antibodies
Type: Primary
Antigen: RELB
Clonality: Monoclonal
Clone: 315206
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR