Anti-NFKB1 Mouse Monoclonal Antibody [clone: 285412]

Supplier: R & D Systems
MAB2697
220020-037EA 577.02 USD
220020-037
Anti-NFKB1 Mouse Monoclonal Antibody [clone: 285412]
Antibodies
Type: Primary
Antigen Symbol: NFKB1
Antigen Name: nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
Antigen Synonyms: EBP-1, KBF1, NF-kB1, NF-kappa-B, NF-kappaB, NFKB-p105, NFKB-p50, NFkappaB, p105, p50
Clonality: Monoclonal
Clone: 285412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR