Anti-TMPRSS11D Mouse Monoclonal Antibody [clone: 337029]

Supplier: R & D Systems
MAB2695
220020-036EA 594.37 USD
220020-036
Anti-TMPRSS11D Mouse Monoclonal Antibody [clone: 337029]
Antibodies
Type: Primary
Antigen: TMPRSS11D
Clonality: Monoclonal
Clone: 337029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR