Anti-COLEC12 Rat Monoclonal Antibody [clone: 388305]

Supplier: R & D Systems
MAB26901
220020-035EA 561.11 USD
220020-035
Anti-COLEC12 Rat Monoclonal Antibody [clone: 388305]
Antibodies
Type: Primary
Antigen: COLEC12
Clonality: Monoclonal
Clone: 388305
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR