Anti-PTPRZ1 Rat Monoclonal Antibody [clone: 279245]

Supplier: R & D Systems
MAB26881
220020-032EA 600.15 USD
220020-032
Anti-PTPRZ1 Rat Monoclonal Antibody [clone: 279245]
Antibodies
Type: Primary
Antigen: PTPRZ1
Clonality: Monoclonal
Clone: 279245
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR