Anti-PTPRZ1 Rat Monoclonal Antibody [clone: 279244]

Supplier: R & D Systems
MAB2688
220020-031EA 633.42 USD
220020-031
Anti-PTPRZ1 Rat Monoclonal Antibody [clone: 279244]
Antibodies
Type: Primary
Antigen: PTPRZ1
Clonality: Monoclonal
Clone: 279244
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR