Anti-PIK3CD Mouse Monoclonal Antibody [clone: 275144]

Supplier: R & D Systems
MAB2687
220020-030EA 594.37 USD
220020-030
Anti-PIK3CD Mouse Monoclonal Antibody [clone: 275144]
Antibodies
Type: Primary
Antigen: PIK3CD
Clonality: Monoclonal
Clone: 275144
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR