Anti-PIK3CB Mouse Monoclonal Antibody [clone: 269020]

Supplier: R & D Systems
MAB2686
220020-029EA 577.02 USD
220020-029
Anti-PIK3CB Mouse Monoclonal Antibody [clone: 269020]
Antibodies
Type: Primary
Antigen: PIK3CB
Clonality: Monoclonal
Clone: 269020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR