Anti-NTF4 Mouse Monoclonal Antibody [clone: 36507]

Supplier: R & D Systems
MAB2681
220020-028EA 763.57 USD
220020-028
Anti-NTF4 Mouse Monoclonal Antibody [clone: 36507]
Antibodies
Type: Primary
Antigen: NTF4
Clonality: Monoclonal
Clone: 36507
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR