Anti-NTF4 Mouse Monoclonal Antibody [clone: 36526]

Supplier: R & D Systems
MAB268
220020-026EA 772.25 USD
220020-026
Anti-NTF4 Mouse Monoclonal Antibody [clone: 36526]
Antibodies
Type: Primary
Antigen: NTF4
Clonality: Monoclonal
Clone: 36526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR