Anti-CREBBP Mouse Monoclonal Antibody [clone: 257003]

Supplier: R & D Systems
MAB2676
220020-024EA 600.15 USD
220020-024
Anti-CREBBP Mouse Monoclonal Antibody [clone: 257003]
Antibodies
Type: Primary
Antigen: CREBBP
Clonality: Monoclonal
Clone: 257003
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR