Anti-NTHL1 Mouse Monoclonal Antibody [clone: 208521]

Supplier: R & D Systems
MAB2675
220020-023EA 577.02 USD
220020-023
Anti-NTHL1 Mouse Monoclonal Antibody [clone: 208521]
Antibodies
Type: Primary
Antigen: NTHL1
Clonality: Monoclonal
Clone: 208521
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR