Anti-JUN Mouse Monoclonal Antibody [clone: 320831]

Supplier: R & D Systems
MAB2670
220020-022EA 600.15 USD
220020-022
Anti-JUN Mouse Monoclonal Antibody [clone: 320831]
Antibodies
Type: Primary
Antigen: JUN
Clonality: Monoclonal
Clone: 320831
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR