Anti-YWHAZ Mouse Monoclonal Antibody [clone: 818515]

Supplier: R & D Systems
MAB2669
102885-486EA 594.37 USD
102885-486
Anti-YWHAZ Mouse Monoclonal Antibody [clone: 818515]
Antibodies
Simple Western

Type: Primary
Antigen: YWHAZ
Clonality: Monoclonal
Clone: 818515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR