Anti-BGN Mouse Monoclonal Antibody [clone: 303203]

Supplier: R & D Systems
MAB2667
220020-019EA 594.37 USD
220020-019
Anti-BGN Mouse Monoclonal Antibody [clone: 303203]
Antibodies
Type: Primary
Antigen: BGN
Clonality: Monoclonal
Clone: 303203
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR