Anti-CDCP1 Mouse Monoclonal Antibody [clone: 309121]

Supplier: R & D Systems
MAB26662
220020-018EA 577.02 USD
220020-018
Anti-CDCP1 Mouse Monoclonal Antibody [clone: 309121]
Antibodies
Type: Primary
Antigen: CDCP1
Clonality: Monoclonal
Clone: 309121
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR