Anti-CDCP1 Mouse Monoclonal Antibody [clone: 309112]

Supplier: R & D Systems
MAB26661
220020-017EA 577.02 USD
220020-017
Anti-CDCP1 Mouse Monoclonal Antibody [clone: 309112]
Antibodies
Type: Primary
Antigen: CDCP1
Clonality: Monoclonal
Clone: 309112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR