Anti-LAIR2 Mouse Monoclonal Antibody [clone: 319701]

Supplier: R & D Systems
MAB2665
220020-015EA 623.29 USD
220020-015
Anti-LAIR2 Mouse Monoclonal Antibody [clone: 319701]
Antibodies
Type: Primary
Antigen: LAIR2
Clonality: Monoclonal
Clone: 319701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR