Anti-LAIR1 Mouse Monoclonal Antibody [clone: 342219]

Supplier: R & D Systems
MAB2664
220020-014EA 655.11 USD
220020-014
Anti-LAIR1 Mouse Monoclonal Antibody [clone: 342219]
Antibodies
Type: Primary
Antigen: LAIR1
Clonality: Monoclonal
Clone: 342219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR