Anti-TNFRSF1A-associated Via Death Domain Mouse Monoclonal Antibody [clone: 313203]

Supplier: R & D Systems
MAB2658
220020-009EA 561.11 USD
220020-009
Anti-TNFRSF1A-associated Via Death Domain Mouse Monoclonal Antibody [clone: 313203]
Antibodies
Type: Primary
Antigen: TNFRSF1A-associated Via Death Domain
Clonality: Monoclonal
Clone: 313203
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR