Anti-SHBG Mouse Monoclonal Antibody [clone: 336711]

Supplier: R & D Systems
MAB2656
220020-007EA 584.25 USD
220020-007
Anti-SHBG Mouse Monoclonal Antibody [clone: 336711]
Antibodies
Type: Primary
Antigen: SHBG
Clonality: Monoclonal
Clone: 336711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR