Anti-CHIL1 Rat Monoclonal Antibody [clone: 321924]

Supplier: R & D Systems
MAB2649
220020-005EA 633.42 USD
220020-005
Anti-CHIL1 Rat Monoclonal Antibody [clone: 321924]
Antibodies
Type: Primary
Antigen: CHIL1
Clonality: Monoclonal
Clone: 321924
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR