Anti-PGF Rat Monoclonal Antibody [clone: 358905]

Supplier: R & D Systems
MAB2642
102885-482EA 594.37 USD
102885-482
Anti-PGF Rat Monoclonal Antibody [clone: 358905]
Antibodies
Type: Primary
Antigen: PGF
Clonality: Monoclonal
Clone: 358905
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR