Anti-CD300A Rat Monoclonal Antibody [clone: 232612]

Supplier: R & D Systems
MAB2640
220020-004EA 594.37 USD
220020-004
Anti-CD300A Rat Monoclonal Antibody [clone: 232612]
Antibodies
Type: Primary
Antigen: CD300A
Clonality: Monoclonal
Clone: 232612
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR