Anti-LOXL2 Mouse Monoclonal Antibody [clone: 262418]

Supplier: R & D Systems
MAB2639
220020-002EA 633.42 USD
220020-002
Anti-LOXL2 Mouse Monoclonal Antibody [clone: 262418]
Antibodies
Type: Primary
Antigen Symbol: LOXL2
Antigen Name: lysyl oxidase-like 2
Antigen Synonyms: LOR2, WS9-14
Clonality: Monoclonal
Clone: 262418
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR