Anti-CTS3 Rat Monoclonal Antibody [clone: 264013]

Supplier: R & D Systems
MAB2638
220020-001EA 763.57 USD
220020-001
Anti-CTS3 Rat Monoclonal Antibody [clone: 264013]
Antibodies
Type: Primary
Antigen: CTS3
Clonality: Monoclonal
Clone: 264013
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR