Anti-LAYN Mouse Monoclonal Antibody [clone: 328002]

Supplier: R & D Systems
MAB26371
220020-000EA 772.25 USD
220020-000
Anti-LAYN Mouse Monoclonal Antibody [clone: 328002]
Antibodies
Type: Primary
Antigen: LAYN
Clonality: Monoclonal
Clone: 328002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR