Anti-LAYN Mouse Monoclonal Antibody [clone: 328024]

Supplier: R & D Systems
MAB2637
220019-999EA 600.15 USD
220019-999
Anti-LAYN Mouse Monoclonal Antibody [clone: 328024]
Antibodies
Type: Primary
Antigen: LAYN
Clonality: Monoclonal
Clone: 328024
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR