Anti-GAS1 Rat Monoclonal Antibody [clone: 309325]

Supplier: R & D Systems
MAB2636
220019-998EA 772.25 USD
220019-998
Anti-GAS1 Rat Monoclonal Antibody [clone: 309325]
Antibodies
Type: Primary
Antigen Symbol: GAS1
Antigen Name: growth arrest-specific 1
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 309325
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR