Anti-IL1R2 Mouse Monoclonal Antibody [clone: 34117]

Supplier: R & D Systems
MAB2631
220019-996EA 763.57 USD
220019-996
Anti-IL1R2 Mouse Monoclonal Antibody [clone: 34117]
Antibodies
Type: Primary
Antigen: IL1R2
Clonality: Monoclonal
Clone: 34117
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR