Anti-IL1R2 Mouse Monoclonal Antibody [clone: 32437]

Supplier: R & D Systems
MAB263
220019-995EA 763.57 USD
220019-995
Anti-IL1R2 Mouse Monoclonal Antibody [clone: 32437]
Antibodies
Type: Primary
Antigen: IL1R2
Clonality: Monoclonal
Clone: 32437
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR