Anti-FGF23 Rat Monoclonal Antibody [clone: 283507]

Supplier: R & D Systems
MAB26291
220019-994EA 600.15 USD
220019-994
Anti-FGF23 Rat Monoclonal Antibody [clone: 283507]
Antibodies
Type: Primary
Antigen Symbol: FGF23
Antigen Name: fibroblast growth factor 23
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 283507
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR