Anti-FGF23 Rat Monoclonal Antibody [clone: 283511]

Supplier: R & D Systems
MAB2629
220019-993EA 600.15 USD
220019-993
Anti-FGF23 Rat Monoclonal Antibody [clone: 283511]
Antibodies
Type: Primary
Antigen Symbol: FGF23
Antigen Name: fibroblast growth factor 23
Antigen Synonyms: ADHR, FGFN, HPDR2, HYPF, PHPTC
Clonality: Monoclonal
Clone: 283511
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR