Anti-MSI1 Mouse Monoclonal Antibody [clone: 282613]

Supplier: R & D Systems
MAB2628
220019-992EA 566.89 USD
220019-992
Anti-MSI1 Mouse Monoclonal Antibody [clone: 282613]
Antibodies
Type: Primary
Antigen: MSI1
Clonality: Monoclonal
Clone: 282613
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR