Anti-KLK14 Mouse Monoclonal Antibody [clone: 337903]

Supplier: R & D Systems
MAB2626
220019-990EA 577.02 USD
220019-990
Anti-KLK14 Mouse Monoclonal Antibody [clone: 337903]
Antibodies
Type: Primary
Antigen: KLK14
Clonality: Monoclonal
Clone: 337903
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR