Anti-KLK13 Mouse Monoclonal Antibody [clone: 341419]

Supplier: R & D Systems
MAB2625
220019-989EA 772.25 USD
220019-989
Anti-KLK13 Mouse Monoclonal Antibody [clone: 341419]
Antibodies
Type: Primary
Antigen: KLK13
Clonality: Monoclonal
Clone: 341419
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR