Anti-KLK7 Mouse Monoclonal Antibody [clone: 333931]

Supplier: R & D Systems
MAB2624
220019-988EA 772.25 USD
220019-988
Anti-KLK7 Mouse Monoclonal Antibody [clone: 333931]
Antibodies
Type: Primary
Antigen: KLK7
Clonality: Monoclonal
Clone: 333931
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR