Anti-OSTN Rat Monoclonal Antibody [clone: 311417]

Supplier: R & D Systems
MAB2620
220019-986EA 633.42 USD
220019-986
Anti-OSTN Rat Monoclonal Antibody [clone: 311417]
Antibodies
Type: Primary
Antigen: OSTN
Clonality: Monoclonal
Clone: 311417
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR