Anti-ITGB2 Rat Monoclonal Antibody [clone: 313903]

Supplier: R & D Systems
MAB2618
220019-984EA 577.02 USD
220019-984
Anti-ITGB2 Rat Monoclonal Antibody [clone: 313903]
Antibodies
Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone: 313903
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR