Anti-CLEC4A2 Rat Monoclonal Antibody [clone: 320511]

Supplier: R & D Systems
MAB2617
220019-983EA 594.37 USD
220019-983
Anti-CLEC4A2 Rat Monoclonal Antibody [clone: 320511]
Antibodies
Type: Primary
Antigen: CLEC4A2
Clonality: Monoclonal
Clone: 320511
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR