Anti-SPON2 Mouse Monoclonal Antibody [clone: 331620]

Supplier: R & D Systems
MAB2609
220019-978EA 561.11 USD
220019-978
Anti-SPON2 Mouse Monoclonal Antibody [clone: 331620]
Antibodies
Type: Primary
Antigen: SPON2
Clonality: Monoclonal
Clone: 331620
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR