Anti-HAPLN1 Mouse Monoclonal Antibody [clone: 316119]

Supplier: R & D Systems
MAB2608
220019-977EA 594.37 USD
220019-977
Anti-HAPLN1 Mouse Monoclonal Antibody [clone: 316119]
Antibodies
Type: Primary
Antigen Symbol: HAPLN1
Antigen Name: hyaluronan and proteoglycan link protein 1
Antigen Synonyms: CRT1, CRTL1
Clonality: Monoclonal
Clone: 316119
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR