Anti-GPC5 Mouse Monoclonal Antibody [clone: 297716]

Supplier: R & D Systems
MAB2607
220019-976EA 594.37 USD
220019-976
Anti-GPC5 Mouse Monoclonal Antibody [clone: 297716]
Antibodies
Type: Primary
Antigen Symbol: GPC5
Antigen Name: glypican 5
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 297716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR