Anti-GATA4 Mouse Monoclonal Antibody [clone: 532020]

Supplier: R & D Systems
MAB2606
220019-975EA 600.15 USD
220019-975
Anti-GATA4 Mouse Monoclonal Antibody [clone: 532020]
Antibodies
Type: Primary
Antigen Symbol: GATA4
Antigen Name: GATA binding protein 4
Antigen Synonyms: ASD2, TACHD, TOF, VSD1
Clonality: Monoclonal
Clone: 532020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR