Anti-FGF23 Mouse Monoclonal Antibody [clone: 275802]

Supplier: R & D Systems
MAB2604
220019-973EA 594.37 USD
220019-973
Anti-FGF23 Mouse Monoclonal Antibody [clone: 275802]
Antibodies
Type: Primary
Antigen Symbol: FGF23
Antigen Name: fibroblast growth factor 23
Antigen Synonyms: ADHR, FGFN, HPDR2, HYPF, PHPTC
Clonality: Monoclonal
Clone: 275802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR