Anti-CTF1 Mouse Monoclonal Antibody [clone: 89228]

Supplier: R & D Systems
MAB260
220019-970EA 772.25 USD
220019-970
Anti-CTF1 Mouse Monoclonal Antibody [clone: 89228]
Antibodies
Type: Primary
Antigen: CTF1
Clonality: Monoclonal
Clone: 89228
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR