Anti-CD4 Mouse Monoclonal Antibody [clone: 372717]

Supplier: R & D Systems
MAB2597
220019-968EA 561.11 USD
220019-968
Anti-CD4 Mouse Monoclonal Antibody [clone: 372717]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 372717
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR