Anti-PLG Mouse Monoclonal Antibody [clone: 270412]

Supplier: R & D Systems
MAB2596
220019-967EA 600.15 USD
220019-967
Anti-PLG Mouse Monoclonal Antibody [clone: 270412]
Antibodies
Type: Primary
Antigen: PLG
Clonality: Monoclonal
Clone: 270412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR