Anti-VEGFA Rat Monoclonal Antibody [clone: 128313]

Supplier: R & D Systems
MAB2595
220019-966EA 577.02 USD
220019-966
Anti-VEGFA Rat Monoclonal Antibody [clone: 128313]
Antibodies
Type: Primary
Antigen: VEGFA
Clonality: Monoclonal
Clone: 128313
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR