Anti-ULBP1 Rat Monoclonal Antibody [clone: 237104]

Supplier: R & D Systems
MAB2588
220019-961EA 594.37 USD
220019-961
Anti-ULBP1 Rat Monoclonal Antibody [clone: 237104]
Antibodies
Type: Primary
Antigen: ULBP1
Clonality: Monoclonal
Clone: 237104
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR